MilliporeSigma
  • miR-214 Attenuates Aortic Valve Calcification by Regulating Osteogenic Differentiation of Valvular Interstitial Cells.
miR-214 Attenuates Aortic Valve Calcification by Regulating Osteogenic Differentiation of Valvular Interstitial Cells.
Molecular therapy. Nucleic acids (2020-12-01)
Ning Li, Yifan Bai, Guangwei Zhou, Ye Ma, Mengwei Tan, Fan Qiao, Xin Li, Ming Shen, Xiaowei Song, Xianxian Zhao, Xiaohong Liu, Zhiyun Xu
ABSTRACT
Calcific aortic valve disease (CAVD) is a common heart valve disease in aging populations, and aberrant osteogenic differentiation of valvular interstitial cells (VICs) plays a critical role in the pathogenesis of ectopic ossification of the aortic valve. miR-214 has been validated to be involved in the osteogenesis process. Here, we aim to investigate the role and mechanism of miR-214 in CAVD progression. miR-214 expression was significantly downregulated in CAVD aortic valve leaflets, accompanied by upregulation of osteogenic markers. Overexpression of miR-214 suppressed osteogenic differentiation of VICs, while silencing the expression of miR-214 promoted this function. miR-214 directly targeted ATF4 and Sp7 to modulate osteoblastic differentiation of VICs, which was proved by dual luciferase reporter assay and rescue experiment. miR-214 knockout rats exhibited higher mean transvalvular velocity and gradient. The expression of osteogenic markers in aortic valve leaflets of miR-214 knockout rats was upregulated compared to that of the wild-type group. Taken together, our study showed that miR-214 inhibited aortic valve calcification via regulating osteogenic differentiation of VICs by directly targeting ATF4 and Sp7, indicating that miR-214 may act as a profound candidate of targeting therapy for CAVD.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATF4
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sp7