MilliporeSigma
  • Stabilization of Black Phosphorus by Sonication-Assisted Simultaneous Exfoliation and Functionalization.
Stabilization of Black Phosphorus by Sonication-Assisted Simultaneous Exfoliation and Functionalization.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-10-03)
Maart van Druenen, Timothy Collins, Fionán Davitt, Jessica Doherty, Gillian Collins, Zdeněk Sofer, Justin D Holmes
ABSTRACT
Black phosphorus (BP) has extraordinary properties, but its ambient instability remains a critical challenge. Functionalization has been employed to overcome the sensitivity of BP to ambient conditions while preserving its properties. Herein, a simultaneous exfoliation-functionalization process is reported that functionalizes BP flakes during exfoliation and thus provides increased protection, which can be attributed to minimal exposure of the flakes to ambient oxygen and water. A tetrabutylammonium salt was employed for intercalation of BP, resulting in the formation of flakes with large lateral dimensions. The addition of an aryl iodide or an aryl iodonium salt to the exfoliation solvent creates a scalable strategy for the production of functionalized few-layer BP flakes. The ambient stability of functionalized BP was prolonged to a period of one week, as revealed by STEM, AFM, and X-ray photoelectron spectroscopy.
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%