MilliporeSigma
  • Diosmetin Mitigates Cognitive and Memory Impairment Provoked by Chronic Unpredictable Mild Stress in Mice.
Diosmetin Mitigates Cognitive and Memory Impairment Provoked by Chronic Unpredictable Mild Stress in Mice.
Evidence-based complementary and alternative medicine : eCAM (2021-01-09)
Elham Saghaei, Shakiba Nasiri Boroujeni, Parvin Safavi, Zeinab Borjian Boroujeni, Elham Bijad
ABSTRACT
In the present experimental study, male NMRI mice were exposed to chronic unpredictable mild stress (CUMS) paradigm for 35 days. Diosmetin (at doses of 10, 20, and 40 mg/kg. i.p.) or diosmetin solvent (normal saline + DMSO, 1 ml/kg; i.p.) was administered 30 min before stress induction. After 28 days, memory and cognitive performance were assessed by shuttle box and novel object recognition tests. Finally, antioxidant capacity (FRAP) and malondialdehyde (MDA) level of serum and brain, and serum corticosterone level were evaluated. Behavioral tests showed that CUMS significantly reduced the secondary latency in passive avoidance memory test and diagnosis index in novel object recognition test compared to the control group (P < 0.001), whereas treatment with diosmetin (20 and 40 mg/kg) significantly improved memory performance in the two tests (P < 0.001). In addition, diosmetin (40 mg/kg) could pronouncedly suppress increase in serum corticosterone levels, reduction in antioxidant capacity, and production of excess MDA caused by CUMS compared to the control group (P < 0.01, P < 0.001, and P < 0.001, respectively). Chronic stress can impair memory and cognition and treatment with diosmetin can partly improve this disorder in male mice by increasing the antioxidant capacity of brain tissue and serum and improving serum corticosterone levels.
Sigma-Aldrich
Diosmetin
Pricing and availability is not currently available.