MilliporeSigma
  • Brown adipose tissue uptake of triglyceride-rich lipoprotein-derived fatty acids in diabetic or obese mice under different temperature conditions.
Brown adipose tissue uptake of triglyceride-rich lipoprotein-derived fatty acids in diabetic or obese mice under different temperature conditions.
EJNMMI research (2020-10-22)
Andreas Paulus, Natascha Drude, Wouter van Marken Lichtenbelt, Felix M Mottaghy, Matthias Bauwens
ABSTRACT
In vivo imaging of glucose analogue 2-deoxy-2-[18F]fluoro-D-glucose ([18F]FDG) via positron emission tomography (PET) is the current gold standard to visualize and assess brown adipose tissue (BAT) activity. However, glucose metabolism is only a part of the metabolic activity of BAT. [18F]FDG-PET has been shown in clinical trials to often fail to visualize BAT under insulin-resistant conditions associated with aging and weight gain. We employed a novel developed triglyceride-based tracer to visualize BATs metabolic activity under different temperature conditions as well as under diabetic and obese conditions in preclinical models. [18F]BDP-TG-chylomicron-like particles visualized BAT in control, streptozocin-induced diabetes and obese mice. Increased BAT tracer uptake was found in control mice acutely exposed to cold but not in cold-acclimated animals. Diabetes did not remove BAT tracer uptake, but did limit BAT tracer uptake to levels of control mice housed at 21 °C. In obese animals, BAT tracer uptake was significantly reduced, although the stimulating effect of cold exposure could still be noted. BAT was visualized in control, diabetic and obese conditions. Streptozocin-induced diabetes, but not obesity, inhibited the stimulatory effect of cold exposure.
Sigma-Aldrich
Sodium p-toluenesulfonate, 95%
Pricing and availability is not currently available.
Triolein, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.