MilliporeSigma
  • Synthesis and anti-mycobacterial evaluation of some pyrazine-2-carboxylic acid hydrazide derivatives.
Synthesis and anti-mycobacterial evaluation of some pyrazine-2-carboxylic acid hydrazide derivatives.
European journal of medicinal chemistry (2010-05-22)
Mohamed Abdel-Aziz, Hamdy M Abdel-Rahman
ABSTRACT
A series of pyrazine-2-carboxylic acid hydrazide derivatives were synthesized and screened for their activity against Mycobacterium tuberculosis. The results show that pyrazine-2-carboxylic acid hydrazide-hydrazone derivatives 3a-l were less active than pyrazinamide. In contrast, the N(4)-ethyl-N(1)-pyrazinoyl-thiosemicarbazide 4 showed the highest activity against M. tuberculosis H(37)Rv (IC(90) = 16.87 microg/mL). Details of the structure-activity and structure-cytotoxicity relationships are discussed.
Sigma-Aldrich
Pyrazinecarboxylic acid, 99%