MilliporeSigma
  • Liposome-mediated delivery of challenging chemicals to aid environmental assessment of Bioaccumulative (B) and Toxic (T) properties.
Liposome-mediated delivery of challenging chemicals to aid environmental assessment of Bioaccumulative (B) and Toxic (T) properties.
Scientific reports (2020-06-18)
Mafalda Castro, Dennis Lindqvist
ABSTRACT
Standard aquatic toxicity tests of chemicals are often limited by the chemicals' water solubility. Liposomes have been widely used in the pharmaceutical industry to overcome poor pharmacokinetics and biodistribution. In this work, liposomes were synthesized and used in an ecotoxicological context, as a tool to assure stable dosing of technically challenging chemicals to zooplankton. Three chemicals with distinctly different characteristics were successfully incorporated into the liposomes: Tetrabromobisphenol A (TBBPA, log Kow 5.9, pKa1 7.5, pKa2 8.5), chlorinated paraffin CP-52 (log Kow 8-12) and perfluorooctanoic acid (PFOA, pKa 2.8). The size, production yield and stability over time was similar for all blank and chemical-loaded liposomes, except for when the liposomes were loaded with 10 or 100 mg g-1 PFOA. PFOA increased the size and decreased the production yield and stability of the liposomes. Daphnia magna were exposed to blank and chemical-loaded liposomes in 48 hour incubation experiments. A dose-dependent increase in body burden in D. magna and increased immobilization (LD50 = 7.6 ng CPs per individual) was observed. This confirms not only the ingestion of the liposomes but also the successful internalization of chemicals. This study shows that liposomes can be a reliable alternative to aid the study of aquatic toxicity of challenging chemicals.
Sigma-Aldrich
3,3′,5,5′-Tetrabromobisphenol A, 97%