MilliporeSigma
  • Cadmium toxicity mediated by the inhibition of SLC2A4 expression in human proximal Tubule cells.
Cadmium toxicity mediated by the inhibition of SLC2A4 expression in human proximal Tubule cells.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-12-19)
Jin-Yong Lee, Maki Tokumoto, Masahiko Satoh
ABSTRACT
Cadmium (Cd) is an environmental contaminant that causes renal toxicity. We have previously demonstrated that Cd induces renal toxicity by altering transcriptional activities. In this study, we show that Cd markedly inhibited the activity of transcription factor MEF2A in HK-2 human proximal tubule cells, which generated significant cytotoxicity in the cells. This reduction in the nuclear levels of MEF2A protein may be involved in the Cd-induced inhibition of MEF2A activity. We also demonstrate that one of the glucose transporters, GLUT4, was downregulated not only by Cd treatment but also by MEF2A knockdown. Knockdown of SLC2A4, encoding GLUT4, eliminated both cell viability and Cd toxicity. Cd treatment or SLC2A4 deficiency reduced the cellular concentration of glucose. Therefore, the suppression of SLC2A4 expression, which mediates the reduction in cellular glucose, is involved in Cd toxicity. The Cd toxicity induced by the reduction in GLUT4 may be associated with a reduction of cellular ATP levels in HK-2 cells. The levels of Slc2a4 mRNA in the kidney of mice exposed to Cd for 6 or 12 months were significantly lower than those in the control group. These results demonstrate that Cd exerts its cytotoxicity through the suppression in SLC2A4 expression and the subsequent inhibition of MEF2A transcriptional activity. Cd-induced suppression of SLC2A4 expression also reduces cellular ATP levels, partly by reducing glucose levels. This study suggests that the glucose transporter plays an important role in the renal toxicity of Cd, and provides a crucial breakthrough in our understanding of the mechanism of Cd toxicity.
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC2A2
Sigma-Aldrich
MISSION® esiRNA, targeting human MEF2A