MilliporeSigma
  • Protein kinase G phosphorylates the Alzheimer's disease-associated tau protein at distinct Ser/Thr sites.
Protein kinase G phosphorylates the Alzheimer's disease-associated tau protein at distinct Ser/Thr sites.
BioFactors (Oxford, England) (2021-01-21)
Giulia Montalto, Francesca Caudano, Laura Sturla, Santina Bruzzone, Annalisa Salis, Gianluca Damonte, Jos Prickaerts, Ernesto Fedele, Roberta Ricciarelli
ABSTRACT
Intraneuronal accumulation of hyperphosphorylated tau is a pathological hallmark of several neurodegenerative disorders, including Alzheimer's disease. Phosphorylation plays a crucial role in modulating the tau-microtubule interaction and the ability of the protein to aggregate, but despite efforts during the past decades, the real identity of the kynases involved in vivo remains uncertain. Here, for the first time, we demonstrate that the cGMP-dependent protein kinase G (PKG) phosphorylates tau in both in vitro and in vivo models. More intriguingly, we provide evidence that PKG phosphorylates tau at Ser214 but not at Ser202, a condition that could reduce the pathological aggregation of the protein shifting tau from a pro-aggregant to a neuroprotective anti-aggregant conformation.
Sigma-Aldrich
MISSION® esiRNA, targeting human PRKG1