MilliporeSigma
  • Efficient microwave-assisted prenylation of pinostrobin and biological evaluation of its derivatives as antitumor agents.
Efficient microwave-assisted prenylation of pinostrobin and biological evaluation of its derivatives as antitumor agents.
Bioorganic & medicinal chemistry letters (2010-03-12)
Hadi Poerwono, Shigeru Sasaki, Yoshiyuki Hattori, Kimio Higashiyama
ABSTRACT
Pinostrobin (5-hydroxy-7-methoxyflavanone) obtained in relatively large amounts from fingerroot (Boesenbergia pandurata) was converted to its C-6 and C-8 prenylated derivatives. The Mitsunobu reaction, europium(III)-catalyzed Claisen-Cope rearrangement, and Claisen reaction coupled with cross-metathesis were used as the key steps. Using a sealed-vessel microwave reactor, the Mitsunobu and Claisen/Cope reactions occurred smoothly with short reaction times and in satisfactory yields. The target compounds and five new intermediary substances showed cytotoxic activity toward SK-BR-3, MCF-7, PC-3, and Colo-320DM human tumor cell lines, and all of them had significantly lower IC(50) (microM) values than pinostrobin.