MilliporeSigma
  • The flavonoid 6-hydroxyflavone prevention of cisplatin-induced nephrotoxicity.
The flavonoid 6-hydroxyflavone prevention of cisplatin-induced nephrotoxicity.
Histology and histopathology (2020-09-11)
Zia Ud Din, Syed Umer Farooq, Muhammad Shahid, Osama Alghamdi, Nawwaf Al-Hamoudi, Fahim Vohra, Tariq Abduljabbar
ABSTRACT
In this study, the flavonoid, 6-hydroxyflavone was investigated for its renal protective activity in the cisplatin rat model of nephrotoxicity. Male Sprague-Dawley rats weighing 200-250 g were included in the study. 6-Hydroxyflavone was daily administered at 25 and 50 mg/kg (i.p.), while ascorbic acid was used as a positive control and injected (i.p.) at 50 mg/kg for 15 days. The nephrotoxicity was evoked with a single cisplatin injection at 7.5 mg/kg on the tenth day of treatment. The renal function and levels of oxidative stress markers were assessed. Each tissue slide of different groups was observed under a compound microscope attached with a digital camera. Cisplatin significantly decreased the overall body weight with an increase in serum creatinine and urea and production of severe histopathological and oxidative stress in the kidneys. The daily treatment with 6-hydroxyflavone significantly attenuated the cisplatin associated detrimental changes in the body weight, and serum levels of creatinine and urea at both 25 mg/kg (P<0.05) and 50 mg/kg (P<0.01). The 6-hydroxyflavone treatment also preserved the renal histoarchitecture from the toxicological influence of cisplatin as evident from a significant reduction in the severity of histopathological changes in the renal tissues. Moreover, 6-hydroxyflavone also reduced the cisplatin-induced lipid peroxidation and corrected the renal antioxidant status. A similar protective effect was observed with the positive control, ascorbic acid (50 mg/kg). These findings show that the flavonoid 6-hydroxyflavone has potential nephroprotective properties and can be used for the management of chemotherapy associated renal disturbances.
Sigma-Aldrich
6-Hydroxyflavone, 98%
Pricing and availability is not currently available.