MilliporeSigma
  • Synthesis of phosphatidylinositol 3-kinase (PI3K) inhibitory analogues of the sponge meroterpenoid liphagal.
Synthesis of phosphatidylinositol 3-kinase (PI3K) inhibitory analogues of the sponge meroterpenoid liphagal.
Journal of medicinal chemistry (2010-12-03)
Alban R Pereira, Wendy K Strangman, Frederic Marion, Larry Feldberg, Deborah Roll, Robert Mallon, Irwin Hollander, Raymond J Andersen
ABSTRACT
Analogues of the sponge meroterpenoid liphagal have been synthesized and evaluated for inhibition of PI3Kα and PI3Kγ as part of a program aimed at developing new isoform-selective PI3K inhibitors. One of the analogues, compound 24, with IC₅₀ values of 66 nM against PI3Kα and 1840 nM against PI3Kγ, representing a 27-fold preference for PI3Kα, exhibited enhanced chemical stability and modestly enhanced potency and selectivity compared with the natural product liphagal.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)