MilliporeSigma
  • Exogenous pancreatic kininogenase protects against renal fibrosis in rat model of unilateral ureteral obstruction.
Exogenous pancreatic kininogenase protects against renal fibrosis in rat model of unilateral ureteral obstruction.
Acta pharmacologica Sinica (2020-04-18)
Ji-Zhe Jin, Hui-Ying Li, Jian Jin, Shang-Guo Piao, Xiong-Hu Shen, Yan-Ling Wu, Jia-Chong Xu, Long-Ye Zhang, Yu-Ji Jiang, Hai-Lan Zheng, Ying-Shun Jin, Sheng Cui, Kang Luo, Yi Quan, Can Li
ABSTRACT
Tissue kallikrein has protective function against various types of injury. In this study, we investigated whether exogenous pancreatic kininogenase (PK) conferred renoprotection in a rat model of unilateral ureteral obstruction (UUO) and H2O2-treated HK-2 cells in vitro. SD rats were subjected to UUO surgery, then PK (7.2 U/g per day, ip) was administered for 7 or 14 days. After the treatment, rats were euthanized; the obstructed kidneys were harvested for further examination. We found that PK administration significantly attenuated interstitial inflammation and fibrosis, and downregulated the expression of proinflammatory (MCP-1, TLR-2, and OPN) and profibrotic (TGF-β1 and CTGF) cytokines in obstructed kidney. UUO-induced oxidative stress, closely associated with excessive apoptotic cell death and autophagy via PI3K/AKT/FoxO1a signaling, which were abolished by PK administration. We further showed that PK administration increased the expression of bradykinin receptors 1 and 2 (B1R and B2R) mRNA and the production of NO and cAMP in kidney tissues. Coadministration with either B1R antagonist (des-Arg9-[Leu8]-bradykinin) or B2R antagonist (icatibant) abrogated the renoprotective effects of PK, and reduced the levels of NO and cAMP in obstructed kidney. In H2O2-treated HK-2 cells, addition of PK (6 pg/mL) significantly decreased ROS production, regulated the expression of oxidant and antioxidant enzymes, suppressed the expression of TGF-β1 and MCP-1, and inhibited cell apoptosis. Our data demonstrate that PK treatment protects against the progression of renal fibrosis in obstructed kidneys.
Sigma-Aldrich
Lys-(des-Arg9, Leu8)-Bradykinin trifluoroacetate salt, ≥95%
Sigma-Aldrich
HOE 140, ≥94%