MilliporeSigma
  • 4-(Dimethylamino)pyridine N-Oxide-Catalyzed Macrolactamization Using 2-Methyl-6-nitrobenzoic Anhydride in the Synthesis of the Depsipeptidic Analogue of FE399.
4-(Dimethylamino)pyridine N-Oxide-Catalyzed Macrolactamization Using 2-Methyl-6-nitrobenzoic Anhydride in the Synthesis of the Depsipeptidic Analogue of FE399.
ACS omega (2021-02-16)
Takayuki Tonoi, Takehiko Inohana, Ryo Kawahara, Teruyuki Sato, Miyuki Ikeda, Miku Akutsu, Takatsugu Murata, Isamu Shiina
ABSTRACT
A depsipeptidic analogue of FE399 was efficiently synthesized mainly through macrolactamization using 2-methyl-6-nitrobenzoic anhydride (MNBA), and a detailed investigation of the desired 16-membered macrolactam core of FE399 was performed. It was determined that the combination of MNBA and a catalytic amount of 4-(dimethylamino)pyridine N-oxide exhibits much higher activity than that of conventionally used coupling reagents such as hexafluorophosphate azabenzotriazole tetramethyl uronium and benzotriazol-1-yl-oxytripyrrolidinophosphonium hexafluorophosphate.
Sigma-Aldrich
2-Methyl-6-nitrobenzoic anhydride, 97%