MilliporeSigma
  • Evidences of CTLA-4 and PD-1 Blocking Agents-Induced Cardiotoxicity in Cellular and Preclinical Models.
Evidences of CTLA-4 and PD-1 Blocking Agents-Induced Cardiotoxicity in Cellular and Preclinical Models.
Journal of personalized medicine (2020-10-23)
Vincenzo Quagliariello, Margherita Passariello, Domenica Rea, Antonio Barbieri, Martina Iovine, Annamaria Bonelli, Antonietta Caronna, Gerardo Botti, Claudia De Lorenzo, Nicola Maurea
ABSTRACT
Several strategies based on immune checkpoint inhibitors (ICIs) have been developed for cancer therapy, opening to advantages in cancer outcomes. However, several ICI-induced side effects have emerged in these patients, especially a rare but clinically significant cardiotoxicity with high rate of mortality. We studied the cytotoxic and pro-inflammatory properties of Ipilimumab and Nivolumab, the underlying pathways and cytokine storm involved. Co-cultures of human cardiomyocytes and lymphocytes were exposed to Ipilimumab or Nivolumab; cell viability and expression of leukotrienes, NLRP3, MyD88, and p65/NF-kB were performed. C57 mice were treated with Ipilimumab (15 mg/kg); analysis of fractional shortening, ejection fraction, radial and longitudinal strain were made before and after treatments through 2D-echocardiography. Expression of NLRP3, MyD88, p65/NF-kB, and 12 cytokines were analyzed in murine myocardium. Nivolumab and Ipilimumab exert effective anticancer, but also significant cardiotoxic effects in co-cultures of lymphocytes and tumor or cardiac cells. Both ICIs increased NLRP3, MyD88, and p65/NF-kB expression compared to untreated cells, however, the most pro-inflammatory and cardiotoxic effects were seen after exposure to Ipilimumab. Mice treated with Ipilimumab showed a significant decrease in fractional shortening and radial strain with respect to untreated mice, coupled with a significant increase in myocardial expression of NLRP3, MyD88, and several interleukins. Nivolumab and Ipilimumab exert cytotoxic effects mediated by the NLRP3/IL-1β and MyD88 pathways, leading to pro-inflammatory cytokine storm in heart tissue.
Sigma-Aldrich
Leukotriene B4, ~100 μg/mL in ethanol, ≥97%