MilliporeSigma
  • ETS1 Suppresses Tumorigenesis of Human Breast Cancer via Trans-Activation of Canonical Tumor Suppressor Genes.
ETS1 Suppresses Tumorigenesis of Human Breast Cancer via Trans-Activation of Canonical Tumor Suppressor Genes.
Frontiers in oncology (2020-06-02)
Gi-Cheon Kim, Choong-Gu Lee, Ravi Verma, Dipayan Rudra, Taemook Kim, Keunsoo Kang, Jong Hee Nam, Young Kim, Sin-Hyeog Im, Ho-Keun Kwon
ABSTRACT
ETS1 has shown dichotomous roles as an oncogene and a tumor suppressor gene in diverse cancers, but its functionality in breast cancer tumorigenesis still remains unclear. We utilized the Cancer Genome Atlas (TCGA) database to analyze comprehensive functions of ETS1 in human breast cancer (BRCA) patients by investigating its expression patterns and methylation status in relation to clinical prognosis. ETS1 expression was significantly diminished by hyper-methylation of the ETS1 promoter region in specimens from BRCA patients compared to a healthy control group. Moreover, ETS1 high BRCA patients showed better prognosis and longer survival compared to ETS1 low BRCA patients. Consistent with clinical evidence, comparative transcriptome analysis combined with CRISPR/Cas9 or shRNA based perturbation of ETS1 expression revealed direct as well as indirect mechanisms of ETS1 that hinder tumorigenesis of BRCA cells. Taken together, our study enlightens a novel function of ETS1 as a tumor suppressor in breast cancer cells.
Sigma-Aldrich
Ionomycin, Free Acid, Streptomyces conglobatus, Ionomycin, Free Acid, Streptomyces conglobatus, CAS 56092-81-0, is a highly specific for divalent cations and more effective than A23187 (Cat. No. 100105) as a mobile ion carrier for Ca2+.
Sigma-Aldrich
Phorbol-12-myristate-13-acetate, Phorbol-12-myristate-13-acetate, CAS 16561-29-8, is the most common phorbol ester. Activates PKC at nanomolar concentrations.