MilliporeSigma
Bacterial chemolithoautotrophy via manganese oxidation.
Nature (2020-07-17)
Hang Yu, Jared R Leadbetter
ABSTRACT
Manganese is one of the most abundant elements on Earth. The oxidation of manganese has long been theorized1-yet has not been demonstrated2-4-to fuel the growth of chemolithoautotrophic microorganisms. Here we refine an enrichment culture that exhibits exponential growth dependent on Mn(II) oxidation to a co-culture of two microbial species. Oxidation required viable bacteria at permissive temperatures, which resulted in the generation of small nodules of manganese oxide with which the cells associated. The majority member of the culture-which we designate 'Candidatus Manganitrophus noduliformans'-is affiliated to the phylum Nitrospirae (also known as Nitrospirota), but is distantly related to known species of Nitrospira and Leptospirillum. We isolated the minority member, a betaproteobacterium that does not oxidize Mn(II) alone, and designate it Ramlibacter lithotrophicus. Stable-isotope probing revealed 13CO2 fixation into cellular biomass that was dependent upon Mn(II) oxidation. Transcriptomic analysis revealed candidate pathways for coupling extracellular manganese oxidation to aerobic energy conservation and autotrophic CO2 fixation. These findings expand the known diversity of inorganic metabolisms that support life, and complete a biogeochemical energy cycle for manganese5,6 that may interface with other major global elemental cycles.
Sigma-Aldrich
Sodium nitrate-15N, ≥98 atom % 15N, ≥99% (CP)