MilliporeSigma
  • Evaluation of a semi-automatic isoelectric focusing method for apolipoprotein E phenotyping.
Evaluation of a semi-automatic isoelectric focusing method for apolipoprotein E phenotyping.
Practical laboratory medicine (2020-01-08)
Randa Bittar, Alain Carrié, Georges Nouadje, Corinne Cherfils, Valérie Fesel-Fouquier, Laurence Barbot-Trystram, Philippe Giral, Dominique Bonnefont-Rousselot
ABSTRACT
A qualitative, semi-automatized method for apolipoprotein E (apoE) phenotyping by isoelectric focusing method has been evaluated on 40 serum samples from patients previously genotyped for apoE, especially as regards concordance with genotyping, but also repeatability and reproducibility of the method, and sample storage. Total concordance with genotyping and good precision criteria, together with its practicability and requirement of a little sample volume, lead to conclude to the usefulness of this method to help clinicians in the diagnosis of dyslipidemic and neurodegenerative diseases.
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder