MilliporeSigma
  • An engineered antibody binds a distinct epitope and is a potent inhibitor of murine and human VISTA.
An engineered antibody binds a distinct epitope and is a potent inhibitor of murine and human VISTA.
Scientific reports (2020-09-18)
Nishant Mehta, Sainiteesh Maddineni, Ryan L Kelly, Robert B Lee, Sean A Hunter, John L Silberstein, R Andres Parra Sperberg, Caitlyn L Miller, Amanda Rabe, Louai Labanieh, Jennifer R Cochran
ABSTRACT
V-domain immunoglobulin (Ig) suppressor of T cell activation (VISTA) is an immune checkpoint that maintains peripheral T cell quiescence and inhibits anti-tumor immune responses. VISTA functions by dampening the interaction between myeloid cells and T cells, orthogonal to PD-1 and other checkpoints of the tumor-T cell signaling axis. Here, we report the use of yeast surface display to engineer an anti-VISTA antibody that binds with high affinity to mouse, human, and cynomolgus monkey VISTA. Our anti-VISTA antibody (SG7) inhibits VISTA function and blocks purported interactions with both PSGL-1 and VSIG3 proteins. SG7 binds a unique epitope on the surface of VISTA, which partially overlaps with other clinically relevant antibodies. As a monotherapy, and to a greater extent as a combination with anti-PD1, SG7 slows tumor growth in multiple syngeneic mouse models. SG7 is a promising clinical candidate that can be tested in fully immunocompetent mouse models and its binding epitope can be used for future campaigns to develop species cross-reactive inhibitors of VISTA.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Roche
Liberase TL Research Grade, lyophilized, suitable for tissue processing, optimum pH 7.4
Roche
DNase I, grade II, from bovine pancreas