MilliporeSigma
  • Potential Biomarkers of the Turnover, Mineralization, and Volume Classification: Results Using NMR Metabolomics in Hemodialysis Patients.
Potential Biomarkers of the Turnover, Mineralization, and Volume Classification: Results Using NMR Metabolomics in Hemodialysis Patients.
JBMR plus (2020-07-16)
Aline L Baptista, Kallyandra Padilha, Pamella A Malagrino, Gabriela Venturini, Ana Cm Zeri, Luciene M Dos Reis, Janaina S Martins, Vanda Jorgetti, Alexandre C Pereira, Silvia M Titan, Rosa Ma Moyses
ABSTRACT
Bone biopsy is still the gold standard to assess bone turnover (T), mineralization (M), and volume (V) in CKD patients, and serum biomarkers are not able to replace histomorphometry. Recently, metabolomics has emerged as a new technique that could allow for the identification of new biomarkers useful for disease diagnosis or for the understanding of pathophysiologic mechanisms, but it has never been assessed in the chronic kidney disease-mineral and bone disorder (CKD-MBD) scenario. In this study, we investigated the association between serum metabolites and the bone TMV classification in patients with end-stage renal disease by using serum NMR spectroscopy and bone biopsy of 49 hemodialysis patients from a single center in Brazil. High T was identified in 21 patients and was associated with higher levels of dimethylsulfone, glycine, citrate, and N-acetylornithine. The receiver-operating characteristic curve for the combination of PTH and these metabolites provided an area under the receiver-operating characteristic curve (AUC) of 0.86 (0.76 to 0.97). Abnormal M was identified in 30 patients and was associated with lower ethanol. The AUC for age, diabetes mellitus, and ethanol was 0.83 (0.71 to 0.96). Low V was identified in 17 patients and was associated with lower carnitine. The association of age, phosphate, and carnitine provided an AUC of 0.83 (0.70 to 0.96). Although differences among the curves by adding selected metabolites to traditional models were not statistically significant, the accuracy of the diagnosis according to the TMV classification seemed to be improved. This is the first study to evaluate the TMV classification system in relation to the serum metabolome assessed by NMR spectroscopy, showing that selected metabolites may help in the evaluation of bone phenotypes in CKD-MBD. © 2020 The Authors. JBMR Plus published by Wiley Periodicals, Inc. on behalf of American Society for Bone and Mineral Research.
Product Description
Supelco
3-(Trimethylsilyl)propionic-2,2,3,3-d4 acid sodium salt, 98 atom % D