MilliporeSigma
  • A founder mutation in the PLPBP gene in families from Saguenay-Lac-St-Jean region affected by a pyridoxine-dependent epilepsy.
A founder mutation in the PLPBP gene in families from Saguenay-Lac-St-Jean region affected by a pyridoxine-dependent epilepsy.
JIMD reports (2021-05-13)
Maitou Pal, Baiba Lace, Yvan Labrie, Nathalie Laflamme, Nadie Rioux, Samarth Thonta Setty, Marc-Andre Dugas, Louise Gosselin, Arnaud Droit, Nicolas Chrestian, Serge Rivest
ABSTRACT
Pyridoxine-dependent epilepsy (PDE) is a relatively rare subgroup of epileptic disorders. They generally present in infancy as an early onset epileptic encephalopathy or seizures, refractory to standard treatments, with rapid and variable responses to vitamin B6 treatment. Whole exome sequencing of three unrelated families identified homozygous pathogenic mutation c.370_373del, p.Asp124fs in PLPBP gene in five persons. Haplotype analysis showed a single shared profile for the affected persons and their parents, leading to a hypothesis about founder effect of the mutation in Saguenay-Lac-St-Jean region of French Canadians. All affected probands also shared one single mitochondrial haplotype T2b3 and two rare variations in the mitochondrial genome m.801A>G and m.5166A>G suggesting that a single individual female introduced PLPBP mutation c.370_373del, p.Asp124fs in Quebec. The mutation p.Asp124fs causes a severe disease phenotype with delayed myelination and cortical/subcortical brain atrophy. The most noteworthy radiological finding in this Quebec founder mutation is the presence of the temporal cysts that can be used as a marker of the disease. Also, both patients, who are alive, had a history of prenatal supplements taken by their mothers as antiemetic medication with high doses of pyridoxine. In the context of suspected PDE in patients with neonatal refractory seizures, treatment with pyridoxine and/or Pyridoxal-5-phophate has to be started immediately and continued until the results of genetic analysis received. Even with early appropriate treatment, neurological outcome of our patient is still poor.
Sigma-Aldrich
Anti-PROSC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Pricing and availability is not currently available.