MilliporeSigma
  • iTRAQ-based comparative proteomics analysis reveals specific urinary biomarkers for various kidney diseases.
iTRAQ-based comparative proteomics analysis reveals specific urinary biomarkers for various kidney diseases.
Biomarkers in medicine (2020-08-29)
Juan Jin, Jianguang Gong, Li Zhao, Yiwen Li, Yunguang Wang, Qiang He
ABSTRACT
Background: Proteome studies for multiple renal diseases is bare. Methodology & results: Using isobaric tags for relative and absolute quantitation labeling, many differentially expressed proteins (DEPs) were identified in acute kidney injury (AKI), AKI + chronic kidney disease (CKD), diabetic CKD and nondiabetic CKD with or without IgA nephropathy (IgAN). Comparative analysis indicated that 34, 35, 17, 91 and 14 unique DEPs were found in AKI, AKI + CKD, CKD, diabetic CKD and nondiabetic CKD. Compared with nondiabetic CKD with IgAN, 47 unique DEPs were found in that without IgAN. Serum amyloid A1 (SAA1) and hepatocyte growth factor activator were unregulated in AKI and nondiabetic CKD without IgAN, respectively. Regenerating islet-derived protein 3-α (Reg3A) upregulation is associated with AKI and AKI + CKD patients. Conclusion: This research contributes to urinary biomarker discovery from multiple renal diseases.
Sigma-Aldrich
iTRAQ® Reagent - 8PLEX Multiplex Kit
Pricing and availability is not currently available.