MilliporeSigma
  • Comparison of phosphorylation and assembly of photosystem complexes and redox homeostasis in two wheat cultivars with different drought resistance.
Comparison of phosphorylation and assembly of photosystem complexes and redox homeostasis in two wheat cultivars with different drought resistance.
Scientific reports (2017-10-07)
Yang-Er Chen, Jun-Mei Cui, Yan-Qiu Su, Chao-Ming Zhang, Jie Ma, Zhong-Wei Zhang, Ming Yuan, Wen-Juan Liu, Huai-Yu Zhang, Shu Yuan
ABSTRACT
Reversible phosphorylation of proteins and the assembly of thylakoid complexes are the important protective mechanism against environmental stresses in plants. This research was aimed to investigate the different responses of the antioxidant defense system and photosystem II (PSII) to osmotic stress between drought-resistant and drought-susceptible wheat cultivars. Results showed that the decrease in PSII photochemistry and six enzyme activities was observed in drought-susceptible wheat compared with drought-resistant wheat under osmotic stress. In addition, a lower accumulation of reactive oxygen species (ROS) and cell death were found in the resistant wheat compared with the susceptible wheat under osmotic stress. Western blot analysis revealed that osmotic stress led to a remarkable decline in the steady state level of D1 protein in drought-susceptible wheat. However, the CP29 protein was strongly phosphorylated in drought-resistant wheat compared with the susceptible wheat under osmotic stress. Our results also showed that drought-resistant wheat presented higher phosphorylated levels of the light-harvesting complex II (LHCII), D1, and D2 proteins and a more rapid dephosphorylated rate than drought-susceptible wheat under osmotic stress. Furthermore, the PSII-LHCII supercomplexes and LHCII trimers were more rapidly disassembled in drought-susceptible wheat than the drought-resistant wheat under osmotic stress. These findings provide that reversible phosphorylation of thylakoid membrane proteins and assembly of thylakoid membrane complexes play important roles in plant adaptation to environmental stresses.