MilliporeSigma
  • Mitochondrial copper depletion suppresses triple-negative breast cancer in mice.
Mitochondrial copper depletion suppresses triple-negative breast cancer in mice.
Nature biotechnology (2020-10-21)
Liyang Cui, Arvin M Gouw, Edward L LaGory, Shenghao Guo, Nabeel Attarwala, Yao Tang, Ji Qi, Yun-Sheng Chen, Zhou Gao, Kerriann M Casey, Arkadiy A Bazhin, Min Chen, Leeann Hu, Jinghang Xie, Mingxi Fang, Cissy Zhang, Qihua Zhu, Zhiyuan Wang, Amato J Giaccia, Sanjiv Sam Gambhir, Weiping Zhu, Dean W Felsher, Mark D Pegram, Elena A Goun, Anne Le, Jianghong Rao
ABSTRACT
Depletion of mitochondrial copper, which shifts metabolism from respiration to glycolysis and reduces energy production, is known to be effective against cancer types that depend on oxidative phosphorylation. However, existing copper chelators are too toxic or ineffective for cancer treatment. Here we develop a safe, mitochondria-targeted, copper-depleting nanoparticle (CDN) and test it against triple-negative breast cancer (TNBC). We show that CDNs decrease oxygen consumption and oxidative phosphorylation, cause a metabolic switch to glycolysis and reduce ATP production in TNBC cells. This energy deficiency, together with compromised mitochondrial membrane potential and elevated oxidative stress, results in apoptosis. CDNs should be less toxic than existing copper chelators because they favorably deprive copper in the mitochondria in cancer cells instead of systemic depletion. Indeed, we demonstrate low toxicity of CDNs in healthy mice. In three mouse models of TNBC, CDN administration inhibits tumor growth and substantially improves survival. The efficacy and safety of CDNs suggest the potential clinical relevance of this approach.
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit, for cellular ATP determination
Sigma-Aldrich
D-Glucose-13C6, endotoxin tested, 99 atom % 13C