MilliporeSigma
  • Label-free cell-based assay with spectral-domain optical coherence phase microscopy.
Label-free cell-based assay with spectral-domain optical coherence phase microscopy.
Journal of biomedical optics (2014-04-09)
Suho Ryu, Kyung-A Hyun, Jung Heo, Hyo-Il Jung, Chulmin Joo
ABSTRACT
Quantitative measurement of dynamic responses of unstained living cells is of great importance in many applications ranging from investigation of fundamental cellular functions to drug discoveries. Conventional optical methods for label-free cell-based assay examine cellular structural changes proximal to sensor surfaces under external stimuli, but require dedicated nanostructure-patterned substrates for operation. Here, we present a quantitative imaging method, spectral-domain optical coherence phase microscopy (SD-OCPM), as a viable optical platform for label-free cell-based assay. The instrument is based on a low-coherence interferometric microscope that enables quantitative depth-resolved phase measurement of a transparent specimen with high phase stability. We demonstrate SD-OCPM measurement of dynamic responses of human breast cancer cells (MCF-7) to 2-picolinic acid (PA) and histamine.
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Mini Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies