- Prevalence and susceptibility profile of Candida metapsilosis and Candida orthopsilosis: results from population-based surveillance of candidemia in Spain.
Prevalence and susceptibility profile of Candida metapsilosis and Candida orthopsilosis: results from population-based surveillance of candidemia in Spain.
We describe the prevalences and susceptibility profiles of two recently described species, Candida metapsilosis and Candida orthopsilosis, related to Candida parapsilosis in candidemia. The prevalences of these species (1.7% for C. metapsilosis and 1.4% for C. orthopsilosis) are significant. Differences observed in their susceptibility profiles could have therapeutic importance.