MilliporeSigma
Rapid reverse phase-HPLC assay of HMG-CoA reductase activity.
Journal of lipid research (2010-04-27)
Matteo Mozzicafreddo, Massimiliano Cuccioloni, Anna Maria Eleuteri, Mauro Angeletti
ABSTRACT
Radioisotope-based and mass spectrometry coupled to chromatographic techniques are the conventional methods for monitoring HMG-CoA reductase (HMGR) activity. Irrespective of offering adequate sensitivity, these methods are often cumbersome and time-consuming, requiring the handling of radiolabeled chemicals or elaborate ad-hoc derivatizing procedures. We propose a rapid and versatile reverse phase-HPLC method for assaying HMGR activity capable of monitoring the levels of both substrates (HMG-CoA and NADPH) and products (CoA, mevalonate, and NADP(+)) in a single 20 min run with no pretreatment required. The linear dynamic range was 10-26 pmol for HMG-CoA, 7-27 nmol for NADPH, 0.5-40 pmol for CoA and mevalonate, and 2-27 nmol for NADP(+), and limit of detection values were 2.67 pmol, 2.77 nmol, 0.27 pmol, and 1.3 nmol, respectively.
Sigma-Aldrich
HMG-CoA Reductase Assay Kit, sufficient for 30 assays (1 mL), sufficient for 100 assays (200 μL)