MilliporeSigma
  • Zika Virus Persistence in the Central Nervous System and Lymph Nodes of Rhesus Monkeys.
Zika Virus Persistence in the Central Nervous System and Lymph Nodes of Rhesus Monkeys.
Cell (2017-05-02)
Malika Aid, Peter Abbink, Rafael A Larocca, Michael Boyd, Ramya Nityanandam, Ovini Nanayakkara, Amanda J Martinot, Edward T Moseley, Eryn Blass, Erica N Borducchi, Abishek Chandrashekar, Amanda L Brinkman, Katherine Molloy, David Jetton, Lawrence J Tartaglia, Jinyan Liu, Katharine Best, Alan S Perelson, Rafael A De La Barrera, Mark G Lewis, Dan H Barouch
ABSTRACT
Zika virus (ZIKV) is associated with severe neuropathology in neonates as well as Guillain-Barré syndrome and other neurologic disorders in adults. Prolonged viral shedding has been reported in semen, suggesting the presence of anatomic viral reservoirs. Here we show that ZIKV can persist in cerebrospinal fluid (CSF) and lymph nodes (LN) of infected rhesus monkeys for weeks after virus has been cleared from peripheral blood, urine, and mucosal secretions. ZIKV-specific neutralizing antibodies correlated with rapid clearance of virus in peripheral blood but remained undetectable in CSF for the duration of the study. Viral persistence in both CSF and LN correlated with upregulation of mechanistic target of rapamycin (mTOR), proinflammatory, and anti-apoptotic signaling pathways, as well as downregulation of extracellular matrix signaling pathways. These data raise the possibility that persistent or occult neurologic and lymphoid disease may occur following clearance of peripheral virus in ZIKV-infected individuals.
Product Description
Millipore
MILLIPLEX® Non-Human Primate Cytokine Magnetic Bead Panel - Premixed 23 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology in non-human primate serum, plasma and cell culture samples.