MilliporeSigma
  • Treatment with the selective muscarinic agonist talsaclidine decreases cerebrospinal fluid levels of total amyloid beta-peptide in patients with Alzheimer's disease.
Treatment with the selective muscarinic agonist talsaclidine decreases cerebrospinal fluid levels of total amyloid beta-peptide in patients with Alzheimer's disease.
Annals of the New York Academy of Sciences (2001-02-24)
C Hock, A Maddalena, I Heuser, D Naber, W Oertel, H von der Kammer, M Wienrich, A Raschig, M Deng, J H Growdon, R M Nitsch
ABSTRACT
Brain amyloid load in Alzheimer's disease (AD) is, at least in genetic forms, associated with overproduction of amyloid beta-peptides (A beta). Thus, lowering A beta production is a central therapeutic target in AD and may be achieved by modulating such key enzymes of amyloid precursor protein (APP) processing as beta-, gamma-, and alpha-secretase activities. Talsaclidine is a selective muscarinic M1 agonist that stimulates the nonamyloidogenic alpha-secretase pathway in model systems. Talsaclidine was administered double-blind, placebo-controlled, and randomized to 24 AD patients and cerebrospinal fluid (CSF) levels of total A beta were quantitated before and after 4 weeks of drug treatment. We observed that talsaclidine decreases CSF levels of A beta significantly over time within the treatment group (n = 20) by a median of 16% as well as compared to placebo (n = 4) by a median of 27%. We conclude that treatment with selective M1 agonists may reduce A beta production and may thus be further evaluated as a potential amyloid-lowering therapy of AD.