MilliporeSigma
  • A selective inhibitor of the Polo-box domain of Polo-like kinase 1 identified by virtual screening.
A selective inhibitor of the Polo-box domain of Polo-like kinase 1 identified by virtual screening.
Journal of advanced research (2019-03-23)
Sara Abdelfatah, Angela Berg, Madeleine Böckers, Thomas Efferth
ABSTRACT
Polo-like kinase 1 (PLK1), a member of the Polo-like kinase family, plays an important regulatory role in mitosis and cell cycle progression. PLK1 overexpression is correlated with tumourigenesis and poor prognosis in cancer patients. Therefore, the identification of novel compounds that inhibit PLK1 would provide attractive therapeutic approaches. Although some PLK1 kinase inhibitors have been developed, their application has been limited by off-target effects. PLK1 contains a regulatory domain named the Polo-box domain (PBD), which is characteristic only for the Polo-like kinase family. This domain represents an alternative therapeutic target with higher selectivity for PLK1. In this study, we applied in silico virtual drug screening, fluorescence polarization and microscale thermophoresis to identify new scaffolds targeting the PBD of PLK1. One compound, 3-{[(1R,9S)-3-(naphthalen-2-yl)-6-oxo-7,11-diazatricyclo[7.3.1.02,7]trideca-2,4-dien-11-yl]methyl}benzonitrile (designated compound (1)), out of a total of 30,793 natural product derivatives, inhibited the PLK1 PBD with high selectivity (IC50: 17.9 ± 0.5 µM). This compound inhibited the growth of cultured leukaemia cells (CCRF-CEM and CEM/ADR5000) and arrested the cell cycle in the G2/M phase, which is characteristic for PLK1 inhibitors. Immunofluorescence analyses showed that treatment with compound (1) disrupted spindle formation due to the aberrant localization of PLK1 during the mitotic process, leading to G2/M arrest and ultimately cell death. In conclusion, compound (1) is a selective PLK1 inhibitor that inhibits cancer cell growth. It represents a chemical scaffold for the future synthesis of new selective PLK1 inhibitors for cancer therapy.
Sigma-Aldrich
PLK1 Polo Box Domain active human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Pricing and availability is not currently available.