MilliporeSigma
  • Stratifin accelerates progression of lung adenocarcinoma at an early stage.
Stratifin accelerates progression of lung adenocarcinoma at an early stage.
Molecular cancer (2015-08-01)
Aya Shiba-Ishii, Yunjung Kim, Toshihiro Shiozawa, Shinji Iyama, Kaishi Satomi, Junko Kano, Shingo Sakashita, Yukio Morishita, Masayuki Noguchi
ABSTRACT
Adenocarcinoma in situ (AIS) of the lung has an extremely favorable prognosis. However, early but invasive adenocarcinoma (eIA) sometimes has a fatal outcome. We had previously compared the expression profiles of AIS with those of eIA showing lymph node metastasis or a fatal outcome, and found that stratifin (SFN, 14-3-3 sigma) was a differentially expressed gene related to cell proliferation. Here, we performed an in vivo study to clarify the role of SFN in initiation and progression of lung adenocarcinoma. Suppression of SFN expression in A549 (a human lung adenocarcinoma cell line) by siSFN significantly reduced cell proliferation activity and the S-phase subpopulation. In vivo, tumor development or metastasis to the lung was reduced in shSFN-transfected A549 cells. Moreover, we generated SFN-transgenic mice (Tg-SPC-SFN(+/-)) showing lung-specific expression of human SFN under the control of a tissue-specific enhancer, the SPC promoter. We found that Tg-SPC-SFN(+/-) mice developed lung tumors at a significantly higher rate than control mice after administration of chemical carcinogen, NNK. Interestingly, several Tg-SPC-SFN(+/-) mice developed tumors without NNK. These tumor cells showed high hSFN expression. These results suggest that SFN facilitates lung tumor development and progression. SFN appears to be a novel oncogene with potential as a therapeutic target.