MilliporeSigma
  • Establishing references for gene expression analyses by RT-qPCR in Theobroma cacao tissues.
Establishing references for gene expression analyses by RT-qPCR in Theobroma cacao tissues.
Genetics and molecular research : GMR (2011-11-19)
T T Pinheiro, C G Litholdo, M L Sereno, G A Leal, P S B Albuquerque, A Figueira
ABSTRACT
Lack of continuous progress in Theobroma cacao (Malvaceae) breeding, especially associated with seed quality traits, requires more efficient selection methods based on genomic information. Reverse transcript quantitative PCR (RT-qPCR) has become the method of choice for gene expression analysis, but relative expression analysis requires various reference genes, which must be stable across various biological conditions. We sought suitable reference genes for various tissues of cacao, especially developing seeds. Ten potential reference genes were analyzed for stability at various stages of embryo development, leaves, stems, roots, flowers, and pod epicarp; seven of them were also evaluated in shoot tips treated either with hormones (salicylate; ethefon; methyl-jasmonate) or after inoculation with the fungus Moniliophthora perniciosa (Marasmiaceae sensu lato). For developing embryos, the three most stable genes were actin (ACT), polyubiquitin (PUB), and ribosomal protein L35 (Rpl35). In the analyses of various tissues, the most stable genes were malate dehydrogenase (MDH), glyceraldehyde 3-phosphate dehydrogenase (GAPDH), and acyl-carrier protein B (ACP B). GAPDH, MDH and tubulin (TUB) were the most appropriate for normalization when shoot apexes were treated with hormones, while ACT, TUB and Rpl35 were the most appropriate after inoculation with M. perniciosa. We conclude that for each plant system and biological or ontogenetical condition, there is a need to define suitable reference genes. This is the first report to define reference genes for expression studies in cacao.