MilliporeSigma
  • High MYBL2 expression and transcription regulatory activity is associated with poor overall survival in patients with hepatocellular carcinoma.
High MYBL2 expression and transcription regulatory activity is associated with poor overall survival in patients with hepatocellular carcinoma.
Current research in translational medicine (2017-12-25)
Z Guan, W Cheng, D Huang, A Wei
ABSTRACT
In this study, we aimed to assess the association between MYBL2 expression/transcription regulatory activity (TRA) and overall survival (OS) in patients with primary hepatocellular carcinoma (HCC) and to explore the factors related to B-Myb TRA. Bioinformatic analysis was performed based on data from the cancer genome atlas-liver hepatocellular carcinoma (TCGA-LIHC) and the human protein atlas (HPA). The death group in TCGA-LIHC had significantly higher MYBL2 RNA and exon expression than the censor group. The high MYBL2 RNA and exon expression groups had significantly worse OS (P<0.01). Univariate and multivariate analysis confirmed that high MYBL2 expression was an independent prognostic factor of unfavourable OS (HR=1.591, 95%CI: 1.119-2.262, P=0.01). One hundred and fourteen out of 188 primary HCC cases in TCGA-LIHC had elevated transcription of B-Myb's downstream genes. High B-Myb TRA was associated with poor OS (P=0.013). Elevated expression of MYBL2, LIN9, LIN52 and FOXM1 were related to the higher TRA of B-Myb in HCC. High MYBL2 expression/TRA are associated with inferior OS in patients with primary HCC. Increased expression of MYBL2, LIN9, LIN52 and FOXM1 are related to higher TRA of B-Myb in HCC.