MilliporeSigma
  • Effect of successive increase in alcohol chains on reaction with isocyanates and isothiocyanates.
Effect of successive increase in alcohol chains on reaction with isocyanates and isothiocyanates.
Natural product research (2009-12-17)
Shahnaz Perveen, Arfa Yasmin, Khalid Mohammed Khan
ABSTRACT
The reaction of isocyanates and isothiocyanates with long-chain alcohols, e.g. n-hexanol, n-heptanol and n-octanol, exclusively gave N-aryl-O-alkyl carbamates, while N-aryl-O-alkyl carbamates were formed along with symmetrical 1,3-disubstituted ureas and thioureas when the same reactions were carried out with small-chain alcohols at room temperature without using any solvent.
Sigma-Aldrich
Benzyl isothiocyanate, 98%
Pricing and availability is not currently available.