MilliporeSigma
  • GBA1 deficiency negatively affects physiological α-synuclein tetramers and related multimers.
GBA1 deficiency negatively affects physiological α-synuclein tetramers and related multimers.
Proceedings of the National Academy of Sciences of the United States of America (2018-01-10)
Sangjune Kim, Seung Pil Yun, Saebom Lee, George Essien Umanah, Veera Venkata Ratnam Bandaru, Xiling Yin, Peter Rhee, Senthilkumar S Karuppagounder, Seung-Hwan Kwon, Hojae Lee, Xiaobo Mao, Donghoon Kim, Akhilesh Pandey, Gabsang Lee, Valina L Dawson, Ted M Dawson, Han Seok Ko
ABSTRACT
Accumulating evidence suggests that α-synuclein (α-syn) occurs physiologically as a helically folded tetramer that resists aggregation. However, the mechanisms underlying the regulation of formation of α-syn tetramers are still mostly unknown. Cellular membrane lipids are thought to play an important role in the regulation of α-syn tetramer formation. Since glucocerebrosidase 1 (GBA1) deficiency contributes to the aggregation of α-syn and leads to changes in neuronal glycosphingolipids (GSLs) including gangliosides, we hypothesized that GBA1 deficiency may affect the formation of α-syn tetramers. Here, we show that accumulation of GSLs due to GBA1 deficiency decreases α-syn tetramers and related multimers and increases α-syn monomers in CRISPR-GBA1 knockout (KO) SH-SY5Y cells. Moreover, α-syn tetramers and related multimers are decreased in N370S GBA1 Parkinson's disease (PD) induced pluripotent stem cell (iPSC)-derived human dopaminergic (hDA) neurons and murine neurons carrying the heterozygous L444P GBA1 mutation. Treatment with miglustat to reduce GSL accumulation and overexpression of GBA1 to augment GBA1 activity reverse the destabilization of α-syn tetramers and protect against α-syn preformed fibril-induced toxicity in hDA neurons. Taken together, these studies provide mechanistic insights into how GBA1 regulates the transition from monomeric α-syn to α-syn tetramers and multimers and suggest unique therapeutic opportunities for PD and dementia with Lewy bodies.