MilliporeSigma
  • Effect of frozen storage, different thawing methods and cooking processes on the survival of Salmonella spp. and Escherichia coli O157:H7 in commercially shaped beef patties.
Effect of frozen storage, different thawing methods and cooking processes on the survival of Salmonella spp. and Escherichia coli O157:H7 in commercially shaped beef patties.
Meat science (2014-12-03)
Stavros G Manios, Panagiotis N Skandamis
ABSTRACT
The effect of common handling practices (i.e., freezing, thawing and cooking) of beef patties on the survival of Salmonella spp. and Escherichia coli O157:H7, was evaluated. Inoculated ground beef was stored at -22 °C for 5 and 75 days. After thawing at 4 °C/16 h, 20 °C/12 h, in microwave/22-24 min, or without prior thawing, beef patties (90 g) were shaped and cooked in oven-broiler or in pan-grill to internal temperatures of 60 °C or 71 °C. Cooking in oven-broiler was more effective compared to pan-grill, especially when cooked to 71 °C. Defrosting methods did not affect significantly (P ≥ 0.05) the survival of the pathogens during subsequent cooking. Frozen storage for 75 days enhanced the survival of E. coli O157:H7, as the pathogen survived 3.1 logCFU/g when cooked in oven-broiler at 71 °C. Results may supplement the existing guidelines for the appropriate practices, associated with freezing, thawing and cooking of patties in households or catering services.
Product Description
Millipore
Kovac′s reagent for indoles, suitable for microbiology
Pricing and availability is not currently available.