MilliporeSigma
  • The P2 promoter of the IGF1 gene is a major epigenetic locus for GH responsiveness.
The P2 promoter of the IGF1 gene is a major epigenetic locus for GH responsiveness.
The pharmacogenomics journal (2015-04-15)
M Ouni, M P Belot, A L Castell, D Fradin, P Bougnères
ABSTRACT
Short children using growth hormone (GH) to accelerate their growth respond to this treatment with a variable efficacy. The causes of this individual variability are multifactorial and could involve epigenetics. Quantifying the impact of epigenetic variation on response to treatments is an emerging challenge. Here we show that methylation of a cluster of CGs located within the P2 promoter of the insulin-like growth factor 1 (IGF1) gene, notably CG-137, is inversely closely correlated with the response of growth and circulating IGF1 to GH administration. For example, variability in CG-137 methylation contributes 25% to variance of growth response to GH. Methylation of CGs in the P2 promoter is negatively associated with the increased transcriptional activity of P2 promoter in patients' mononuclear blood cells following GH administration. Our observation indicates that epigenetics is a major determinant of GH signaling (physiology) and of individual responsiveness to GH treatment (pharmacoepigenetics).