MilliporeSigma
  • Inhibition of antibody production in vivo by pre-stimulation of Toll-like receptor 4 before antigen priming is caused by defective B-cell priming and not impairment in antigen presentation.
Inhibition of antibody production in vivo by pre-stimulation of Toll-like receptor 4 before antigen priming is caused by defective B-cell priming and not impairment in antigen presentation.
International immunology (2012-10-19)
Nurlaely Mida Rachmawati, Kenji Fukudome, Naoko Tsuneyoshi, Uleng Bahrun, Hiroki Tsukamoto, Tsutomu Yanagibashi, Yoshinori Nagai, Kiyoshi Takatsu, Shoichiro Ohta, Masao Kimoto
ABSTRACT
Stimulation of Toll-like receptor 4 (TLR4) induces not only innate but also adaptive immune responses, and has been suggested to exert adjuvant effects. Additional to such positive effects, pre-stimulation of TLR4 induces endotoxin tolerance where animals are unresponsive to subsequent lethal challenges with lipopolysaccharide (LPS). We examined the effects of pre-stimulation of TLR4 using an agonistic anti-TLR4 mAb (UT12) on antibody production in vivo. Pre-injection of UT12 prior to both primary and secondary immunization completely inhibited antigen-specific antibody responses. Cellular analysis revealed that the inhibition was not due to impairment of T-cell activation. Accordingly, T-helper activities in UT12 pre-injected mice were not impaired. In contrast, B-cell priming was defective in UT12 pre-injected mice. The observation that the expression of activation markers such as CD69 and CD86 on B cells was blocked by UT12 pre-injection supports this. Interestingly, UT12 pre-injection only showed inhibitory effects at the primary and not the secondary immunization. These results provide important information concerning the regulatory mechanisms of antibody production, especially in endotoxin-tolerant states.
Sigma-Aldrich
IgG3, Lambda from murine myeloma, clone Y5606, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.