MilliporeSigma
  • Antisense oligonucleotide and adjuvant exercise therapy reverse fatigue in old mice with myotonic dystrophy.
Antisense oligonucleotide and adjuvant exercise therapy reverse fatigue in old mice with myotonic dystrophy.
Molecular therapy. Nucleic acids (2021-01-22)
Ningyan Hu, Eunjoo Kim, Layal Antoury, Jia Li, Paloma González-Pérez, Seward B Rutkove, Thurman M Wheeler
ABSTRACT
Patients with myotonic dystrophy type 1 (DM1) identify chronic fatigue as the most debilitating symptom, which manifests in part as prolonged recovery after exercise. Clinical features of DM1 result from pathogenic gain-of-function activity of transcripts containing an expanded microsatellite CUG repeat (CUGexp). In DM1 mice, therapies targeting the CUGexp transcripts correct the molecular phenotype, reverse myotonia, and improve muscle pathology. However, the effect of targeted molecular therapies on fatigue in DM1 is unknown. Here, we use two mouse models of DM1, age-matched wild-type controls, an exercise-activity assay, electrical impedance myography, and therapeutic antisense oligonucleotides (ASOs) to show that exaggerated exercise-induced fatigue progresses with age, is unrelated to muscle fiber size, and persists despite correction of the molecular phenotype for 3 months. In old DM1 mice, ASO treatment combined with an exercise training regimen consisting of treadmill walking 30 min per day 6 days per week for 3 months reverse all measures of fatigue. Exercise training without ASO therapy improves some measures of fatigue without correction of the molecular pathology. Our results highlight a key limitation of ASO monotherapy for this clinically important feature and support the development of moderate-intensity exercise as an adjuvant for targeted molecular therapies of DM1.
Product Description
Roche
Anti-Digoxigenin, from mouse IgG1κ (clone 1.71.256)