MilliporeSigma
  • Expression and characterization of recombinant kurtoxin, an inhibitor of T-type voltage-gated calcium channels.
Expression and characterization of recombinant kurtoxin, an inhibitor of T-type voltage-gated calcium channels.
Biochemical and biophysical research communications (2011-11-19)
Chul Won Lee, Young-Jae Eu, Hye Jung Min, Eun-Mi Cho, Jun-Ho Lee, Ha Hyung Kim, Seung-Yeol Nah, Kenton J Swartz, Jae Il Kim
ABSTRACT
Kurtoxin, a 63-amino acid peptide stabilized by four disulfide bonds, is the first reported peptide inhibitor of T-type voltage-gated calcium channels. Although T-type calcium channels have been implicated in a number of disease states, including epilepsy, chronic pain, hypertension and cancer, the lack of selective inhibitors has slowed progress in understanding their precise roles. Kurtoxin is a potentially valuable tool with which to study T-type calcium channels. However, because of the limited availability of the native protein, little is known about the structure and molecular mechanism of kurtoxin. Here we report the expression of kurtoxin in Escherichia coli and the structural and functional characterization of the recombinant protein. The disulfide bond pairings and secondary structure of recombinant kurtoxin were characterized through enzymatic cleavage, mass analysis and CD spectroscopy. Recombinant kurtoxin almost completely inhibited the T-type calcium channel in a manner identical to the native toxin. The availability of recombinant kurtoxin that is identical to the native toxin should help in the study of T-type calcium channels and enable development of new strategies for producing even more-selective T-type calcium channel inhibitors and for investigating the molecular basis of the toxin-channel interactions.