MilliporeSigma
  • Absence of Severe Acute Respiratory Syndrome Coronavirus 2 Neutralizing Activity in Prepandemic Sera From Individuals With Recent Seasonal Coronavirus Infection.
Absence of Severe Acute Respiratory Syndrome Coronavirus 2 Neutralizing Activity in Prepandemic Sera From Individuals With Recent Seasonal Coronavirus Infection.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2020-12-04)
Daniel Poston, Yiska Weisblum, Helen Wise, Kate Templeton, Sara Jenks, Theodora Hatziioannou, Paul Bieniasz
ABSTRACT
Cross-reactive immune responses elicited by seasonal coronaviruses might affect severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) susceptibility and disease outcomes. We measured neutralizing activity against SARS-CoV-2 in prepandemic sera from patients with prior polymerase chain reaction scan-confirmed seasonal coronavirus infection. Although neutralizing activity against seasonal coronaviruses was detected in nearly all sera, cross-reactive neutralizing activity against SARS-CoV-2 was undetectable.
Sigma-Aldrich
Anti-Coronavirus Group Antigen Antibody, nucleoprotein of OC-43, clone 542-7D, clone 542-7D, Chemicon®, from mouse