MilliporeSigma
  • Evaluation of nine surface disinfectants against Candida auris using a quantitative disk carrier method: EPA SOP-MB-35.
Evaluation of nine surface disinfectants against Candida auris using a quantitative disk carrier method: EPA SOP-MB-35.
Infection control and hospital epidemiology (2020-07-01)
D Joseph Sexton, Rory M Welsh, Meghan L Bentz, Kaitlin Forsberg, Brendan Jackson, Elizabeth L Berkow, Anastasia P Litvintseva
ABSTRACT
We tested 9 disinfectants against Candida auris using the quantitative disk carrier method EPA-MB-35-00: 5 products with hydrogen peroxide or alcohol-based chemistries were effective and 4 quaternary ammonium compound-based products were not. This work supported a FIFRA Section 18 emergency exemption granted by the US Environmental Protection Agency to expand disinfectant guidance for C. auris.
Product Description
Millipore
Dey-Engley neutralizing broth, suitable for microbiology, NutriSelect® Plus
Pricing and availability is not currently available.