MilliporeSigma
  • Role for p38 mitogen-activated protein kinase in platelet aggregation caused by collagen or a thromboxane analogue.
Role for p38 mitogen-activated protein kinase in platelet aggregation caused by collagen or a thromboxane analogue.
The Journal of biological chemistry (1996-03-22)
J Saklatvala, L Rawlinson, R J Waller, S Sarsfield, J C Lee, L F Morton, M J Barnes, R W Farndale
ABSTRACT
p38 mitogen-activated protein kinase (MAPK) was identified in platelets on the basis of (a) its reactivity with antibodies to C-terminal and N-terminal peptides, and (b) its ability to activate MAPK-activated protein kinase-2, which phosphorylates the small heat shock protein, hsp27. p38 MAPK was activated in platelets by collagen fibers, a collagen-related cross-linked peptide, thrombin, or the thromboxane analogue U46619. A highly specific inhibitor of p38 MAPK, a pyridinyl imidazole known as SB203580, inhibited the platelet enzyme in vitro (IC50 approximately 0.5 microM). At similar concentrations it also inhibited agonist-stimulated phosphorylation of hsp27 in platelets, and platelet aggregation and secretion induced by minimal aggregatory concentrations of collagen or U46619, but not thrombin. Inhibition of aggregation was overcome by increasing agonist dose. SB203580 might act by inhibiting thromboxane generation, but this was only inhibited by 10-20% at low agonist concentrations. p38 MAPK provides a crucial signal, which is necessary for aggregation caused by minimal concentrations of collagen fibers or U46619. Thrombin or high doses of these agonists generate signals that bypass the enzyme, or render the enzyme no longer rate-limiting.
Sigma-Aldrich
p38/SAPK2 Inhibitor (SB 203580), The p38/SAPK2 Inhibitor (SB 203580) controls the biological activity of p38/SAPK2. This small molecule/inhibitor is primarily used for Biochemicals applications.
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)