MilliporeSigma
  • Uncoupling protein-2 negatively regulates insulin secretion and is a major link between obesity, beta cell dysfunction, and type 2 diabetes.
Uncoupling protein-2 negatively regulates insulin secretion and is a major link between obesity, beta cell dysfunction, and type 2 diabetes.
Cell (2001-07-07)
C Y Zhang, G Baffy, P Perret, S Krauss, O Peroni, D Grujic, T Hagen, A J Vidal-Puig, O Boss, Y B Kim, X X Zheng, M B Wheeler, G I Shulman, C B Chan, B B Lowell
ABSTRACT
beta cells sense glucose through its metabolism and the resulting increase in ATP, which subsequently stimulates insulin secretion. Uncoupling protein-2 (UCP2) mediates mitochondrial proton leak, decreasing ATP production. In the present study, we assessed UCP2's role in regulating insulin secretion. UCP2-deficient mice had higher islet ATP levels and increased glucose-stimulated insulin secretion, establishing that UCP2 negatively regulates insulin secretion. Of pathophysiologic significance, UCP2 was markedly upregulated in islets of ob/ob mice, a model of obesity-induced diabetes. Importantly, ob/ob mice lacking UCP2 had restored first-phase insulin secretion, increased serum insulin levels, and greatly decreased levels of glycemia. These results establish UCP2 as a key component of beta cell glucose sensing, and as a critical link between obesity, beta cell dysfunction, and type 2 diabetes.
Sigma-Aldrich
Anti-Uncoupling Protein-2 (UCP-2) (144-157) Rabbit pAb, liquid, Calbiochem®
Pricing and availability is not currently available.