MilliporeSigma
  • Urinary Extracellular Vesicles and Salt-Losing Tubulopathies: A Proteomic Approach.
Urinary Extracellular Vesicles and Salt-Losing Tubulopathies: A Proteomic Approach.
Proteomes (2020-05-14)
Francesca Raimondo, Clizia Chinello, Luigi Porcaro, Fulvio Magni, Marina Pitto
ABSTRACT
Renal tubular cells release urinary extracellular vesicles (uEV) that are considered a promising source of molecular markers for renal dysfunction and injury. We investigated uEV proteomes of patients with hereditary salt-losing tubulopathies (SLTs), focusing on those caused by Gitelman and Bartter (BS) syndromes, to provide potential markers for differential diagnosis. Second morning urine was collected from patients with genetically proven SLTs and uEV were isolated by the ultracentrifugation-based protocol. The uEV proteome was run through a diagonal bidimensional electrophoresis (16BAC/SDS-PAGE), to improve hydrophobic protein resolution. Sixteen differential spots from the proteome of two variants (BS2 and BS3) were analysed by nLC-ESI-MS/MS after in-gel tryptic digestion. A total of 167 protein species were identified from 7 BS2 spots and 9 BS3 spot. Most of these proteins were membrane-associated proteins, in particular transmembrane proteins, and were related to typical renal functions. The differential content of some uEV was then validated by immunoblotting. Our work suggests that uEV proteomics represents a promising strategy for the identification of differential SLT proteins. This could play a role in understanding the pathophysiological disease mechanisms and may support the recognition of different syndromes.
Sigma-Aldrich
Anti-SLC12A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution