MilliporeSigma
  • Prevention of adverse electrical and mechanical remodeling with biventricular pacing in a rabbit model of myocardial infarction.
Prevention of adverse electrical and mechanical remodeling with biventricular pacing in a rabbit model of myocardial infarction.
Heart rhythm (2008-01-09)
Samir Saba, Michael A Mathier, Haider Mehdi, Erdal Gursoy, Tong Liu, Bum-Rak Choi, Guy Salama, Barry London
ABSTRACT
Biventricular (BIV) pacing can improve cardiac function in heart failure (HF). This study sought to investigate the mechanisms of benefit of BIV pacing using a rabbit model of myocardial infarction (MI). New Zealand White rabbits were divided into 4 groups (sham-operated [C], MI with no pacing [MI], MI with right ventricular pacing [MI+RV], and MI with BIV pacing [MI+BIV]) and underwent serial electrocardiograms and echocardiograms. At 4 weeks, hearts were excised and tissue was extracted from various areas of the left ventricle (LV). Four weeks after coronary ligation, BIV pacing prevented systolic and diastolic dilation of the LV as well as the reduction in its fractional shortening, restored the QRS width and the rate-dependent QT intervals to their baseline values, and prevented the decline of the ether-a-go-go (Erg) protein levels. This prevention of remodeling was not documented in the MI+RV groups. In this rabbit model of BIV pacing and MI, we show prevention of adverse mechanical and electrical remodeling of the heart. These changes may underlie some of the benefits seen with BIV pacing in HF patients with more severe LV dysfunction.
Sigma-Aldrich
Anti-Potassium Channel HERG Antibody, CT, Chemicon®, from rabbit