MilliporeSigma
  • Characterization of Citrobacter sp. line 328 as a source of Vi for a Vi-CRM(197) glycoconjugate vaccine against Salmonella Typhi.
Characterization of Citrobacter sp. line 328 as a source of Vi for a Vi-CRM(197) glycoconjugate vaccine against Salmonella Typhi.
Journal of infection in developing countries (2013-01-02)
Simona Rondini, Francesca Micoli, Luisa Lanzilao, Ivan Pisoni, Vito Di Cioccio, Allan J Saul, Laura B Martin
ABSTRACT
Salmonella enterica serovar Typhi is the causative agent of typhoid fever with over 22 million cases and over 200,000 deaths reported annually. A vaccine is much needed for use in young children and the Novartis Vaccines Institute for Global Health (NVGH) is developing a conjugate vaccine which targets S. Typhi Vi capsular polysaccharide. Here we describe a method suitable for industrial scale production of the Vi antigen based on expression by a Citrobacter line. We optimized the production of Vi by selecting a suitable Citrobacter strain (Citrobacter 328) that yields high and stable expression of Vi in chemically defined medium under industrial-scale fermentation conditions. Vi-CRM197 made using Vi from Citrobacter 328 elicited high anti-Vi antibody levels in mice and rabbits. Citrobacter 328 is a suitable strain for production of Vi for conjugate anti-Typhi vaccines. Being a BSL-1 organism, which grows in defined medium and stably produces high yields of Vi, it offers excellent potential for safe production of inexpensive vaccines for populations at risk of typhoid fever.
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution