MilliporeSigma
  • Analysis of bovine immunoglobulin G in milk, colostrum and dietary supplements: a review.
Analysis of bovine immunoglobulin G in milk, colostrum and dietary supplements: a review.
Analytical and bioanalytical chemistry (2007-06-21)
Leyton W Gapper, David E J Copestake, Don E Otter, Harvey E Indyk
ABSTRACT
The immunoprotective properties of bovine milk immunoglobulin G (IgG) have led to a recent proliferation of nutritional products incorporating this protein. It has therefore become critical that reliable analytical techniques for the measurement of the IgG content in such products are available. This literature review surveys current methods of analysis for IgG, including separation-based or immuno-based concentration analysis. The review also discusses nutraceutical applications, regulatory issues, stability of IgG and the significance of primary reference material in IgG analysis.
Sigma-Aldrich
IgG from bovine serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder