MilliporeSigma
Combined immunodeficiency caused by a novel homozygous NFKB1 mutation.
The Journal of allergy and clinical immunology (2020-09-28)
Amarilla B Mandola, Nigel Sharfe, Zahra Nagdi, Harjit Dadi, Linda Vong, Daniele Merico, Bo Ngan, Brenda Reid, Chaim M Roifman
ABSTRACT
Genetic faults in several components of the nuclear factor-κB pathway cause immunodeficiency. Most defects lead to combined immunodeficiency with a range of severity. Heterozygous mutations in NFKB1 were associated with common variable immunodeficiency, however, homozygous mutations have not been described. We studied the molecular basis of combined immunodeficiency in a patient who presented with failure to thrive, persistent EBV viremia and hepatitis, pneumocystis jirovecii pneumonitis, and generalized lymphadenopathy. Whole genome and exome sequencing followed by Sanger confirmation were performed to identify the genetic defect. Molecular and cellular techniques were used to assess the variant impact on the nuclear factor-κB pathway and lymphocyte function. Genetic analysis revealed a novel homozygous mutation in NFKB1, c.2878G>A, p.Gly960Arg (G960R). This affected p105 phosphorylation and p50 formation on antigen and cytokine stimulation, as well as attenuating nuclear signal transmission. As a result, both T- and B-cell maturation and function were perturbed. The number of memory CD4+ T cells were reduced, while CD8+ T cells consisted predominately of expanded differentiated populations. The function of T cells were diminished as shown by reduced responses to mitogens as well as diminished cytokine secretion. B-cell maturation was also affected, with decreased IgD+CD27+ memory B cells while transitional B cells were increased, likely contributing to the reduced ability to produce specific antibodies. Homozygous G960R mutation in NFKB1 leads to a severe clinical presentation of combined immunodeficiency. This was associated with blockade of nuclear factor-κB pathway signaling, resulting in aberrations in T- and B-cell maturation and function.
Sigma-Aldrich
Anti-Giα3 Antibody, serum, Upstate®