MilliporeSigma
  • Hydroxyl radical scavenging by edaravone derivatives: Efficient scavenging by 3-methyl-1-(pyridin-2-yl)-5-pyrazolone with an intramolecular base.
Hydroxyl radical scavenging by edaravone derivatives: Efficient scavenging by 3-methyl-1-(pyridin-2-yl)-5-pyrazolone with an intramolecular base.
Bioorganic & medicinal chemistry letters (2006-09-26)
Hidehiko Nakagawa, Ryo Ohyama, Ayako Kimata, Takayoshi Suzuki, Naoki Miyata
ABSTRACT
We synthesized various 3-methyl-1-phenyl-5-pyrazolone (edaravone) derivatives and evaluated their oxidation potential and hydroxyl radical scavenging activity. It was found 3-methyl-1-(pyridin-2-yl)-5-pyrazolone had a much higher ability to scavenge the radical than did edaravone itself. Its efficient radical scavenging activity was assumed to be due to the increase of its anion form, an active form, by a hydrogen-bonded intramolecular base.
Sigma-Aldrich
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%