MilliporeSigma
  • Floratheasaponins A-C, acylated oleanane-type triterpene oligoglycosides with anti-hyperlipidemic activities from flowers of the tea plant (Camellia sinensis).
Floratheasaponins A-C, acylated oleanane-type triterpene oligoglycosides with anti-hyperlipidemic activities from flowers of the tea plant (Camellia sinensis).
Journal of natural products (2005-09-27)
Masayuki Yoshikawa, Toshio Morikawa, Kaori Yamamoto, Yasuyo Kato, Akifumi Nagatomo, Hisashi Matsuda
ABSTRACT
The methanolic extract and its n-butanol-soluble fraction from the flowers of the tea plant (Camellia sinensis) were found to suppress serum triglyceride elevation in olive oil-treated mice. From the n-butanol-soluble fraction, three new acylated oleanane-type triterpene oligoglycosides, floratheasaponins A-C (1-3), were isolated together with several flavonol glycosides and catechins. The structures of 1-3 were elucidated on the basis of chemical and physicochemical evidence as 21-O-angeloyl-22-O-acetyltheasapogenol B 3-O-[beta-D-galactopyranosyl(1-->2)][beta-D-xylopyranosyl(1-->2)-alpha-L-arabinopyranosyl(1-->3)]-beta-D-glucopyranosiduronic acid, 21,22-di-O-angeloyl-R1-barrigenol 3-O-[beta-D-galactopyranosyl(1-->2)][beta-D-xylopyranosyl(1-->2)-alpha-L-arabinopyranosyl(1-->3)]-beta-D-glucopyranosiduronic acid, and 21-O-angeloyl-22-O-2-methylbutyryl-R1-barrigenol 3-O-[beta-D-galactopyranosyl(1-->2)][beta-D-xylopyranosyl(1-->2)-alpha-L-arabinopyranosyl(1-->3)]-beta-D-glucopyranosiduronic acid, respectively. Floratheasaponins (1-3) showed inhibitory effects on serum triglyceride elevation, with their activities being more potent than those of theasaponins E1 (4) and E2 (5) obtained previously from the seeds of C. sinensis.
Sigma-Aldrich
Clofibrate, liquid
Pricing and availability is not currently available.